Learn More
Nickel Electro™ Stainless Steel Rectangular Electric Boiling Baths
Get optimum boiling results with this rectangular, stainless steel electrically heated boiling bath. Featuring an energy regulator that controls the amount of boiling, this Clifton™ Stainless Steel Rectangular Electric Boiling Bath is ideal for safety.
Brand: Nickel Electro™
Code : 04
Additional Details : Weight : 1.00000kg
Description
- Rectangular, stainless steel electrically heated
- Multi-place baths with fixed temperature for boiling water
- Fitted with concentric ringed lids to vary the hole size
- Features an integral constant level device to maintain constant water level during boiling
- Fitted with a sheathed immersion element and a one shot thermal cut out for safety
Specifications
Constant boiling no control | |
4.54 L | |
Distilling/Concentrating Liquids, Melting Agar and Other Solids, Metallurgical Analysis, Protein Ana | |
1101 ER | |
Rectangular | |
321 x 219 x 90 mm | |
90 mm | |
3.3 kg |
Boiling Bath | |
Simmer to boiling at 100°C | |
Stainless Steel | |
6 x 80 mm Holes | |
2 Years | |
321 mm | |
219 mm | |
230 V |