Learn More
Thermo Scientific™ ClipTip™ Pipette Tip Racks
GREENER_CHOICE
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
Brand:  Thermo Scientific™ 94410819
18.17 GBP valid until 2025-06-30
Use promo code "25076" to get your promotional price.
 View more versions of this product
Product Code. 13236269
  • £28.85 / Pack of 8
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
  • Small Rack: For use with ClipTip 20 and 50 Pipette Tips
  • Medium Rack: For use with ClipTip 200 and 300 Pipette Tips
  • Large Rack: For use with ClipTip 1000 Pipette Tips
Ordering Information:
Racks are supplied without tips and inserts
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ClipTip Empty Rack, Large
Rack
ClipTip
ClipTip Empty Rack
ClipTip 1000 Pipette Tips (94420718)
Product Suggestions
Product Suggestions
Videos
SDS
Documents Product Certifications
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2301510 Certificate TypeCertificate of Compliance Date26/01/2023 Catalog Number94410819
Lot Number2236110 Certificate TypeCertificate of Compliance Date12/01/2023 Catalog Number94410819
Lot Number2234110 Certificate TypeCertificate of Compliance Date29/12/2022 Catalog Number94410819
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ ClipTip™ Pipette Tip Racks > ClipTip Empty Rack, Large