Learn More
Thermo Scientific™ ClipTip™ Pipette Tip Racks
GREENER_CHOICE
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
Brand:  Thermo Scientific™ 94410619
 View more versions of this product
Product Code. 15922021
  • £29.05 / Pack of 8
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Utilize Thermo Scientific™ ClipTip™ Pipette Tip Racks, available in three sizes.
  • Small Rack: For use with ClipTip 20 and 50 Pipette Tips
  • Medium Rack: For use with ClipTip 200 and 300 Pipette Tips
  • Large Rack: For use with ClipTip 1000 and 1250 Pipette Tips
  • CT300 Ext Rack: For use with ClipTip 300 Ext Pipette Tips
Ordering Information:
Racks are supplied without tips and inserts
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ClipTip Empty Rack
ClipTip Empty Rack
ClipTip 300 Ext Pipette Tip
ClipTip