missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLOCK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-126
Additional Details : Weight : 0.00970kg
Description
CLOCK Polyclonal specifically detects CLOCK in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation.Specifications
CLOCK | |
Polyclonal | |
Western Blot 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
O08785 | |
CLOCK | |
Two peptides derived from mouse CLOCK conjugated to albumin. [UniProt# O08785] | |
0.1 mL | |
Chromatin Research, Circadian Rhythm, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
9575 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ChIP Assay | |
Unconjugated | |
Whole antisera with 0.05% Sodium Azide | |
BHLHE8, circadian locomoter output cycles protein kaput, Class E basic helix-loop-helix protein 8, clock (mouse) homolog, clock homolog (mouse), EC 2.3.1.48, hCLOCK, KAT13D, KIAA0334bHLHe8circadian locomoter output cycles kaput protein | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
Human, Mouse | |
Antisera |
For Research Use Only