missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLOCK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£161.98 - £348.80
Specifications
Antigen | CLOCK |
---|---|
Dilution | Western Blot 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
Applications | Western Blot, ChIP Assay |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18796793
|
Novus Biologicals
NB100-126SS |
0.025 mL |
£161.98
25µL |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18767003
|
Novus Biologicals
NB100-126 |
0.1 mL |
£348.80
0.10mL |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CLOCK Polyclonal specifically detects CLOCK in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation.Specifications
CLOCK | |
Western Blot, ChIP Assay | |
Unconjugated | |
Rabbit | |
Chromatin Research, Circadian Rhythm, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
Whole antisera with 0.05% Sodium Azide | |
BHLHE8, circadian locomoter output cycles protein kaput, Class E basic helix-loop-helix protein 8, clock (mouse) homolog, clock homolog (mouse), EC 2.3.1.48, hCLOCK, KAT13D, KIAA0334bHLHe8circadian locomoter output cycles kaput protein | |
CLOCK | |
IgG | |
Unpurified |
Western Blot 1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse | |
O08785 | |
9575 | |
Two peptides derived from mouse CLOCK conjugated to albumin. [UniProt# O08785] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only