Learn More
CLOCK Rabbit, anti-Human, Clone: 1I23, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech 82829-1-RR-100UL
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 18065415
  • £429.23 / 100µL
Estimated Shipment: 23-07-2024
to see stock.
Description
Description
The CLOCK antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human CLOCK. This antibody recognizes human, mouse, rat antigen. The CLOCK antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
CLOCK
1I23
bHLHe8, CLOCK, clock homolog (mouse), hCLOCK, KAT13D, KIAA0334
100 μL
9575
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, Immunohistochemistry, ELISA
800 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa