missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLOCK Rabbit, anti-Human, Clone: 1I23, Proteintech
Rabbit Recombinant Monoclonal Antibody
£153.82 - £429.23
Specifications
Antigen | CLOCK |
---|---|
Clone | 1I23 |
Concentration | 800 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18065415
|
Proteintech
82829-1-RR-100UL |
100 μL |
£429.23
100µL |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18065605
|
Proteintech
82829-1-RR-20UL |
20 μL |
£153.82
20µL |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The CLOCK antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human CLOCK. This antibody recognizes human, mouse, rat antigen. The CLOCK antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
CLOCK | |
800 μg/mL | |
Liquid | |
Human, Mouse, Rat | |
bHLHe8, CLOCK, clock homolog (mouse), hCLOCK, KAT13D, KIAA0334 | |
Primary |
1I23 | |
Western Blot, Immunohistochemistry, ELISA | |
Rabbit | |
9575 | |
IgG | |
Store at -20°C. Stable for one year after shipment. |