Learn More
CLOCK Rabbit, anti-Human, Clone: 1I23, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82829-1-RR-20UL
Additional Details : Weight : 0.00100kg
Description
The CLOCK antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human CLOCK. This antibody recognizes human, mouse, rat antigen. The CLOCK antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
CLOCK | |
1I23 | |
bHLHe8, CLOCK, clock homolog (mouse), hCLOCK, KAT13D, KIAA0334 | |
20 μL | |
9575 | |
Store at -20°C. Stable for one year after shipment. | |
Liquid |
Western Blot, Immunohistochemistry, ELISA | |
800 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse, Rat | |
150 kDa | |
IgG |