missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLOCK Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18094-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that belongs to the basic helix-loop-helix (bHLH) family of transcription factors. Polymorphisms within the encoded protein have been associated with circadian rhythm sleep disorders. A similar protein in mice is a circadian regulator that acts as a transcription factor and forms a heterodimer with aryl hydrocarbon receptor nuclear translocator-like to activate transcription of mouse period 1.Specifications
CLOCK | |
Polyclonal | |
Unconjugated | |
CLOCK | |
bHLHe8, CLOCK, clock homolog (mouse), hCLOCK, KAT13D, KIAA0334 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9575 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15516 | |
CLOCK | |
CLOCK Fusion Protein Ag12826 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |