missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLOCK Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£130.00 - £339.00
Specifications
Antigen | CLOCK |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16838134
|
Proteintech
18094-1-AP-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16828134
|
Proteintech
18094-1-AP-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein that belongs to the basic helix-loop-helix (bHLH) family of transcription factors. Polymorphisms within the encoded protein have been associated with circadian rhythm sleep disorders. A similar protein in mice is a circadian regulator that acts as a transcription factor and forms a heterodimer with aryl hydrocarbon receptor nuclear translocator-like to activate transcription of mouse period 1.Specifications
CLOCK | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O15516 | |
9575 | |
CLOCK Fusion Protein Ag12826 | |
Primary | |
-20°C | |
CLOCK |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
bHLHe8, CLOCK, clock homolog (mouse), hCLOCK, KAT13D, KIAA0334 | |
CLOCK | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |