missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) fluoride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

£71.10 - £449.00

Chemical Identifiers

CAS 10026-17-2
Molecular Formula CoF2
Molecular Weight (g/mol) 96.93
MDL Number MFCD00010941
InChI Key YCYBZKSMUPTWEE-UHFFFAOYSA-L
Synonym cobalt ii fluoride, cobalt fluoride, cobalt difluoride, cobaltous fluoride, hsdb 236, cobalt fluoride pink, cobalt ii fluoride, powder, cobalt ii fluoride, anhydrous, cobalt ii fluoride, ultra dry
PubChem CID 24820
IUPAC Name difluorocobalt
SMILES F[Co]F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11328617
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035740.04
2 g
£71.10
2g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11338617
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035740.09
10 g
£199.00
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15436135
View Documents Promotion Details
Thermo Scientific Alfa Aesar
035740.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

It is used as a catalyst to alloy metals and also used for optical deposition, of which it tremendously improves optical quality, in dental care.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
Specifications

Chemical Identifiers

10026-17-2
96.93
YCYBZKSMUPTWEE-UHFFFAOYSA-L
24820
F[Co]F
CoF2
MFCD00010941
cobalt ii fluoride, cobalt fluoride, cobalt difluoride, cobaltous fluoride, hsdb 236, cobalt fluoride pink, cobalt ii fluoride, powder, cobalt ii fluoride, anhydrous, cobalt ii fluoride, ultra dry
difluorocobalt

Specifications

1400°C
10026-17-2
CoF2
UN2923
cobalt ii fluoride, cobalt fluoride, cobalt difluoride, cobaltous fluoride, hsdb 236, cobalt fluoride pink, cobalt ii fluoride, powder, cobalt ii fluoride, anhydrous, cobalt ii fluoride, ultra dry
YCYBZKSMUPTWEE-UHFFFAOYSA-L
difluorocobalt
24820
99.99%
Ampule
4.46 g/mL
Cobalt(II) fluoride, Ultra dry
1200°C
Powder
MFCD00010941
14,2440
Soluble in water,alcohol and acetone.
F[Co]F
96.93
96.93
Odorless
Hygroscopic
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H351-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause an allergic skin reaction.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301-H314

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 233-061-9

missing translation for 'rtecsNumber' : GG0770000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only