Learn More
Cobalt(II,III) oxide, 99.7% (metals basis), Thermo Scientific Chemicals
For Use With Enamels, Semiconductors, Grinding Wheels
£27.74 - £1607.00
Chemical Identifiers
CAS | 1308-06-1 |
---|---|
Molecular Formula | Co3O4 |
Molecular Weight (g/mol) | 240.80 |
MDL Number | MFCD00010939 |
InChI Key | UBEWDCMIDFGDOO-UHFFFAOYSA-N |
Synonym | oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobaltShow More |
PubChem CID | 11651651 |
SMILES | [O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11349317
|
Thermo Scientific Alfa Aesar
040184.14 |
25 g |
£27.74
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11339317
|
Thermo Scientific Alfa Aesar
040184.22 |
100 g |
£104.52
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11319317
|
Thermo Scientific Alfa Aesar
040184.36 |
500 g |
£370.85
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11329317
|
Thermo Scientific Alfa Aesar
040184.A4 |
2.5 kg |
£1607.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Finds use in enamels, semiconductors, grinding wheels. Used in catalysis, superconductors, ceramics and other fields as an important inorganic material. Also used as catalyst and catalyst carrier, as electrode active material, for glass, porcelain colorants and pigments, as chemical industry oxidant and as a catalyst for organic synthesis, in senior goggles and other filter materials, in temperature and gas sensors, in electronic ceramics, in lithium ion battery electrode materials, magnetic materials, electrochromic devices, enamels, solar energy absorbers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1308-06-1 | |
240.80 | |
UBEWDCMIDFGDOO-UHFFFAOYSA-N | |
11651651 |
Co3O4 | |
MFCD00010939 | |
oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobaltShow More | |
[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3] |
Specifications
900°C (decomposition) | |
Black | |
99.7% (Metals basis) | |
MFCD00010939 | |
Soluble in acids and alkalis. Insoluble in water. | |
[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3] | |
240.80 | |
240.8 | |
6.11 |
1308-06-1 | |
Powder | |
Co3O4 | |
oxocobalt; oxo oxocobaltiooxy cobalt, cobalt ii oxide; cobalt iii oxide, cobalt ii, coo.co2o3, aronis24129, cobaltic tetraoxide,, cobalt ii,iii oxide, powder, <10 mum, cobalt ii oxide; oxo oxocobaltio oxy cobalt, cobalt ii,iii oxide metals basis , 2-6 micron powder, oxidanylidenecobalt; oxidanylidene oxidanylidenecobaltiooxy cobalt | |
UBEWDCMIDFGDOO-UHFFFAOYSA-N | |
oxocobalt;oxo(oxocobaltiooxy)cobalt | |
11651651 | |
Odorless | |
Cobalt(II, III) oxide |
Safety and Handling
GHS H Statement
H351-H317
P201-P202-P261-P281-P285-P304+P341-P308+P313-P342+P311-P501c
H334-H350i
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
missing translation for 'einecsNumber' : 215-157-2
missing translation for 'rtecsNumber' : GG2500000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Cobalt(II,III) oxide
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H350i-May cause cancer by inhalation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.