Learn More
Cobalt(II) nitrate hexahydrate, 99%, pure, Thermo Scientific Chemicals
£38.79 - £172.86
Chemical Identifiers
CAS | 10026-22-9 |
---|---|
Molecular Formula | CoH12N2O12 |
Molecular Weight (g/mol) | 291.03 |
MDL Number | MFCD00149647 |
InChI Key | QGUAJWGNOXCYJF-UHFFFAOYSA-N |
Synonym | cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate |
PubChem CID | 24821 |
ChEBI | CHEBI:86214 |
SMILES | O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10388010
|
Thermo Scientific Acros
213091000 |
100 g | Plastic bottle |
£38.79
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10042620
|
Thermo Scientific Acros
213095000 |
500 g | Plastic bottle |
£104.52
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10567183
|
Thermo Scientific Acros
213090010 |
1 kg | Plastic bottle |
£172.86
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10026-22-9 | |
291.03 | |
QGUAJWGNOXCYJF-UHFFFAOYSA-N | |
24821 | |
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
CoH12N2O12 | |
MFCD00149647 | |
cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate | |
CHEBI:86214 |
Specifications
55.0°C | |
Brown-Red to Pink | |
98.5% min. (Complexometry) | |
Co(NO3)2·6H2O | |
15, 2428 | |
Solubility in water: 1340g/L. Other solubilities: soluble in ethanol and acetone, soluble in most common organic solvents | |
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
291.03 | |
CHEBI:86214 | |
99% | |
1.88 | |
Cobalt(II) nitrate hexahydrate, 99% |
10026-22-9 | |
Adhering Crystalline Powder | |
CoH12N2O12 | |
MFCD00149647 | |
cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate | |
QGUAJWGNOXCYJF-UHFFFAOYSA-N | |
λ²-cobalt(2+) hexahydrate dinitrate | |
24821 | |
291.02 | |
Pure | |
1.8800g/mL |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes serious ey
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remov
GHS Signal Word: Danger
missing translation for 'rtecsNumber' : QU7355500
- Cobalt(II) nitrate hexahydrate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only