Learn More
Cobalt standard solution, for AAS, 1mg/mL Co in 2% HNO3, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 195921000
15.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 29
Additional Details : CAS Number : 10141-05-6 Weight : 0.10200kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
Cobalt standard solution | |
1 mg/ml Co in 2% HNO3 | |
Plastic bottle | |
CoN2O6 | |
UFMZWBIQTDUYBN-UHFFFAOYSA-N | |
cobalt(2+);dinitrate | |
25000 | |
58.93 | |
Miscible with water |
990 to 1010μg/mL of Co(2+) in 2 to 5% HNO3 | |
10141-05-6,7732-18-5 | |
100 mL | |
MFCD00010935 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Co+2] | |
182.941 | |
CHEBI:86209 | |
AAS | |
Solution |
Safety and Handling
- Cobalt atomic absorption standard solution
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H412-Harmful to aquatic life with long lasting effects.
- H290-May be corrosive to metals.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only