Learn More
Cobalt standard solution, for AAS, 1mg/mL Co in 2% HNO3, Thermo Scientific Chemicals
£22.80 - £74.90
Specifications
Chemical Name or Material | Cobalt standard solution |
---|---|
Concentration | 990 to 1010μg/mL of Co(2+) in 2 to 5% HNO3 |
Name Note | 1 mg/ml Co in 2% HNO3 |
CAS | 10141-05-6, 7732-18-5 |
Packaging | Plastic bottle |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10308100
|
Thermo Scientific Acros
195921000 |
100 mL | Plastic bottle |
£22.80
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10246490
|
Thermo Scientific Acros
195925000 |
500 mL | Plastic bottle |
£74.90
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For atomic adsorption spectroscopySpecifications
Cobalt standard solution | |
1 mg/ml Co in 2% HNO3 | |
Plastic bottle | |
MFCD00010935 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Co+2] | |
182.941 | |
CHEBI:86209 | |
AAS | |
Solution |
990 to 1010μg/mL of Co(2+) in 2 to 5% HNO3 | |
10141-05-6, 7732-18-5 | |
CoN2O6 | |
UFMZWBIQTDUYBN-UHFFFAOYSA-N | |
cobalt(2+);dinitrate | |
25000 | |
58.93 | |
Miscible with water |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause cancer by inhalation.
May damage fertility.
Harmful to aquatic life with long lasting effects.
May be corrosive to metals.
GHS P Statement:
Do not handle until all safety precautions have been read and understood.
Use personal protective equipment as required.
Avoid release to the environment.
IF exposed or concerned: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep only in original container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- Cobalt atomic absorption standard solution
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H350i-May cause cancer by inhalation.
- H360F-May damage fertility.
- H412-Harmful to aquatic life with long lasting effects.
- H290-May be corrosive to metals.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only