missing translation for 'onlineSavingsMsg'
Learn More
Cobalt wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals Product Code.: 11301498

Cobalt wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Product Code. 11301498
100 cm
Click to view available options
Quantity:
25 cm
100 cm
This item is not returnable. View return policy

Product Code. 11301498

Brand: Thermo Scientific Alfa Aesar 010946.BY

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]

Specifications

Quantity 100 cm
Percent Purity 99.995%
Grade Puratronic™
Odor Odorless
Weight ≈7.00g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Cobalt wire, 1.0mm (0.04 in.) dia., Annealed

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX25K012 Certificate TypeCertificate of Analysis Date06/01/2025 Catalog Number010946.BU,010946.BY
Lot NumberX25K013 Certificate TypeCertificate of Analysis Date08/11/2024 Catalog Number010946.BU,010946.BY
Lot NumberR17F017 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog Number010946.BU,010946.BY
Lot NumberY04I050 Certificate TypeCertificate of Analysis Date16/11/2022 Catalog Number010946.BU,010946.BY
Lot NumberN16B040 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number010946.BU,010946.BY

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cobalt wire, 1.0mm (0.04in) dia, annealed, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.