Learn More
KARTELL COBAS MIRA™ Multi-Curvette
The COBAS MIRA™ Multi-Curvette is a 12-position polymethyl methacrylate (PMMA) multiple curvette segement for the Cobas Mira™ analyzer.
Brand: KARTELL 0150000
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- 12-position polymethyl methacrylate (PMMA) multiple cuvette segments for the Cobas Mira™; analyzer.
- A high-tech production process and stringent quality control guarantee high reproducibility.
- Available in racks of 15 cuvette segments for devices with continuous throughput or in single segments.
Specifications
Multi-cuvette | |
Whole Blood, Plasma, Serum, Urine | |
15 Segments |