Learn More
KARTELL COBAS MIRA™ Multi-Curvette
The COBAS MIRA™ Multi-Curvette is a 12-position polymethyl methacrylate (PMMA) multiple curvette segement for the Cobas Mira™ analyzer.
£384.00 - £406.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10767524
|
KARTELL
0150100 |
1 Segment |
£384.00
Pack of 450 |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10551393
|
KARTELL
0150000 |
15 Segments |
£406.00
Pack of 450 |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- 12-position polymethyl methacrylate (PMMA) multiple cuvette segments for the Cobas Mira™; analyzer.
- A high-tech production process and stringent quality control guarantee high reproducibility.
- Available in racks of 15 cuvette segments for devices with continuous throughput or in single segments.
Specifications
Multi-cuvette |