missing translation for 'onlineSavingsMsg'
Learn More

Melford Coelenterazine-H

Highly sensitive and specific luniophore used for measuring changes in levels of superoxide and Ca2+ in cells transfected with apoaequorin cDNA.

Brand:  Melford C61500-0.01

 View more versions of this product

Product Code. 30018191

  • £492.00 / 10mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Highly sensitive and specific luniophore used for measuring changes in levels of superoxide and Ca2+ in cells transfected with apoaequorin cDNA.

Exhibits 16-fold greater luminescence intensity as compared to native Coelenterazine.
  • CAS Number: 50909-86-9
  • Molecular Formula: C26H21N3O2
  • Molecular Weight: 407.5
Specifications

Specifications

50909-86-9
C26H21N3O2
10 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers