missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ COEX Cold Extractor

Performs rapid fat extraction directly in the same glass crucibles that are used by the FIWE series for raw fiber analysis. Velp Scientifica™ COEX Cold Extractors can process individual or multiple samples at the same time.

Brand:  Velp Scientifica™ F30530204


Code : NEW

Additional Details : Weight : 19.00000kg

 View more versions of this product

Product Code. 15111296

  • £4105.00 / Each
Estimated Shipment: 18-12-2024
to see stock.
Add to basket

Description

Description

  • No loss - reduced sample remains in the same crucible for analysis
  • Immediate raw fiber extraction after fat extraction here
  • Fatty components collected for easy disposal
  • All reagents can be added through the same channel
Specifications

Specifications

Cold Extractor
Raw Fiber Analysis
300 mm
19 kg
Stainless Steel
1 Ea.
730 mm
230 V 60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers