missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Velp Scientifica™ COEX Cold Extractor
Performs rapid fat extraction directly in the same glass crucibles that are used by the FIWE series for raw fiber analysis. Velp Scientifica™ COEX Cold Extractors can process individual or multiple samples at the same time.
£4105.00
Specifications
Product Type | Cold Extractor |
---|---|
Material | Stainless Steel |
For Use With (Application) | Raw Fiber Analysis |
Quantity | 1 Ea. |
Height (Metric) | 300 mm |
Product Code | Brand | Electrical Requirements | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Electrical Requirements | Price | Quantity & Availability | |||||
15126026
|
Velp Scientifica™
F30520204 |
230 V 50 Hz |
£4105.00
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
15111296
|
Velp Scientifica™
F30530204 |
230 V 60 Hz |
£4105.00
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
Description
- No loss - reduced sample remains in the same crucible for analysis
- Immediate raw fiber extraction after fat extraction here
- Fatty components collected for easy disposal
- All reagents can be added through the same channel
Specifications
Cold Extractor | |
Raw Fiber Analysis | |
300 mm | |
19 kg |
Stainless Steel | |
1 Ea. | |
730 mm |