Learn More
Unity Lab Services™ Cold Storage Equipment Requalification Service with Temperature Mapping
Requalification Services including 1 Temperature Mapping for Cold Storage Equipment (documentation included)
Brand:  Unity Lab Services™ RQTMCOLD
Product Code. 30178677
  • £2763.00 / 1 unit
Explore more special offers
Description
Description
Requalification (RQ): Documented verification that equipment (as installed) is operating as intended according to manufacturers specifications.
  • Verifies important equipment functions and helps ensure that the equipment operates as expected by the manufacturer and conforms to standards and requirements.
  • Key tests include door ajar, power failure, and temperature alerts.
  • Temperature mapping (TM): A mapping study that establishes the temperature distribution within the chamber being mapped and locates hot and cold spots, which provides a view of the equipments performance.
  • Temperature measurements will be performed in the air or with simulated products loaded in a chamber, with a data collection interval configurable to customer needs. The collected data provide essential information, helping ensure all material is correctly stored within its labeled temperature range.
  • TM includes one open door temperature recovery test, one power failure test, minimum and maximum values along with averages, a temperature graph over time, and detailed reports that include raw probe data.
  • Please note that installation and operational qualification, ideally along with preventive maintenance and calibration, should be completed on lab equipment before requalification services can be performed.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cold Storage Equipment
Requalification Service
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Unity Lab Services™ Cold Storage Equipment Requalification Service with Temperature Mapping > Requalification Service