missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Solution, Oakton™

Use this conductivity solution for accurate test results in the field or lab. Oakton™ Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.

Brand:  Cole Parmer 00653-18

 View more versions of this product

Product Code. 11754959

  • £17.85 / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use this conductivity solution for accurate test results in the field or lab. OaktonTM Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.

  • Features temperature corrections printed on the bottle
  • Certified values listed in the tables and are referenced to 25°C
  • Two-year shelf-life on all solutions
  • Conductivity-to-TDS Calibration Values: ppm NaCl: 702.1, ppm 442: 1000, ppm KCl: 744.7
Specifications

Specifications

Cole-Parmer Conductivity Standards
500 mL
Conductivity Standard
1.413 mS (1413 μS)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Statement
  • EUH210-Safety data sheet available on request.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.