Learn More
Conductivity Solution, Oakton™
Use this conductivity solution for accurate test results in the field or lab. Oakton™ Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.
£16.95 - £18.90
Product Code | Brand | Conductivity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Conductivity | Price | Quantity & Availability | |||||
11870120
|
Cole Parmer
00653-23 |
0.023 mS (23 μS) |
£16.95
Each |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11750049
|
Cole Parmer
00653-47 |
0.447 mS (447 μS) |
£16.95
Each |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11754959
|
Cole Parmer
00653-18 |
1.413 mS (1413 μS) |
£16.95
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
11765509
|
Cole Parmer
0065315 |
1.5 mS (1500 μS) |
£18.90
Each |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Use this conductivity solution for accurate test results in the field or lab. OaktonTM Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.
- Features temperature corrections printed on the bottle
- Certified values listed in the tables and are referenced to 25°C
- Two-year shelf-life on all solutions
- Certificates of analysis available separately