missing translation for 'onlineSavingsMsg'
Learn More

Shamrock Color Coded Multi-Purpose Labeling Tape

This Color Coded Multi-Purpose Labeling Tape is designed for organization in laboratories and is removable.

Brand:  Shamrock ST-110-6

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11731626

  • £41.55 / 55m
Estimated Shipment: 12-08-2024
to see stock.

Description

Description

  • Removable labeling tape for organization in laboratories
  • Assorted colors and sizes available
Specifications

Specifications

Removable
Roll
500 in.
1 in. (Core)
Blue
Colored Label Tape
1.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers