missing translation for 'onlineSavingsMsg'
Learn More

Shamrock Color Coded Multi-Purpose Labeling Tape

This Color Coded Multi-Purpose Labeling Tape is designed for organization in laboratories and is removable.

Brand:  Shamrock ST-34-20

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11899070

  • £8.95 / 12m
Estimated Shipment: 19-08-2024
to see stock.

Description

Description

  • Removable labeling tape for organization in laboratories
  • Assorted colors and sizes available
Specifications

Specifications

Removable
Roll
500 in.
1 in. (Core)
Magenta
Colored Label Tape
0.75 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers