Learn More
Epredia™ ColorStack Storage System
Take organization of your slides archives one step further with color-coded drawers.
£306.00 - £410.00
Specifications
Product Type 14-Drawer Cabinet
View More Specs
Products 6
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
12151820
missing translation for 'viewDocuments' Promotion Details
Epredia™
E43/12C/BLUE
Blue
£410.00
Each
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 26-07-2024
to see stock.
12111830
missing translation for 'viewDocuments' Promotion Details
Epredia™
E43/12C/TAN
Tan
£331.00
Each
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 26-07-2024
to see stock.
12141820
missing translation for 'viewDocuments' Promotion Details
Epredia™
E43/12C/AQUA
Aqua This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12191820
missing translation for 'viewDocuments' Promotion Details
Epredia™
E43/12C/ORANGE
Orange This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12131830
missing translation for 'viewDocuments' Promotion Details
Epredia™
E43/12C/YELLOW
Yellow This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
12121830
missing translation for 'viewDocuments' Promotion Details
Epredia™
E43/12C/WHITE
White This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
Description
Description
Includes
  • 14 drawers of one color
  • Each of these storage drawers is 48 x 48 x 14cm and can hold 6,000 slides.
Compatablity
  • Fully compatible with all standard cabinets, bases, and options
  • Colorstack drawers can easily replace drawers in a standard 14-drawer cabinet
  • Special top cover is cream to match the ColorStack 14-Drawer Cabinet
Specifications
Specifications
14-Drawer Cabinet