Learn More
Thermo Scientific™ Columbia Agar with Chocolate Horse Blood
Cultivate, isolate and identify a broad range of microorganisms and fastidious organisms with this multi-purpose nutritious medium containing defibrinated horse blood.
Brand: Thermo Scientific™ PB0124A
13.51 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Cultivate, isolate and identify a broad range of microorganisms and fastidious organisms, using Thermo Scientific™ Columbia Agar with Chocolate Horse Blood. Rapid production of large colonies, clearly defined zones of haemolysis and good colonial differentiation is promoted by the addition of defibrinated horse blood.
Traditionally, blood agar bases have been either casein hydrolysate or meat infusion media. The advantage of the first lies in the rapid production of large colonies, and of the second in clearly defined zones of haemolysis and good colonial differentiation. Columbia Agar Base combines the virtues of both these types of media to give an improved all-round performance, showing versatility and superior performance in several applications.Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.

Specifications
Brown, opaque | |
Columbia Agar with Chocolate Horse Blood | |
United Kingdom | |
19.5 g ± 1.0 g | |
10/Pk. |
2°C to 10°C | |
90mm Monoplate | |
Agar | |
Flow Wrapped - Single Layer |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4500342 | Certificate of Analysis | 31/03/2025 | PB0124A |
4500345 | Certificate of Analysis | 31/03/2025 | PB0124A |
4499860 | Certificate of Analysis | 27/03/2025 | PB0124A |
4500062 | Certificate of Analysis | 26/03/2025 | PB0124A |
4500065 | Certificate of Analysis | 25/03/2025 | PB0124A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.