Learn More
Lenz Laborglasintrumente Columns Acc. to Hempel with Removable Glass Jacket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05117201
Additional Details : Weight : 0.50000kg
Description
- Columns acc. to Hempel
- With removable glass jacket
- For packings like Raschig rings or glass beads
- With NS socket and NS cone
- With removable glass jacket
- Made of DURAN™ tubing
Specifications
Removable glass jacket | |
200 mm | |
NS 14/23 |