Learn More
Lenz Laborglasintrumente Columns Acc. to Hempel with Removable Glass Jacket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
£98.00 - £159.00
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10719411
|
Lenz Laborglasintrumente
05117201 |
200 mm |
£98.00
Each |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10000551
|
Lenz Laborglasintrumente
05117504 |
500 mm |
£123.00
Each |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10010551
|
Lenz Laborglasintrumente
05117804 |
800 mm |
£159.00
Each |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10498402
|
Lenz Laborglasintrumente
05117301 |
300 mm |
£104.00
Each |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Columns acc. to Hempel
- With removable glass jacket
- For packings like Raschig rings or glass beads
- With NS socket and NS cone
- With removable glass jacket
- Made of DURAN™ tubing
Specifications
Removable glass jacket |