Learn More
B Braun™ Combi-Stopper Closing Cones
B Braun™ Combi-Stopper Closing Cones are suitable for sterile closings of IV and syringe assemblies
Brand:  B Braun™ 4495152
 View more versions of this product
Product Code. 10712393
  • £120.00 / Pack of 100
Edge
Explore more special offers
Description
Description
  • Luer Lock fitting male and female
  • Made of PE (Polyethylene)
  • Sterile closing of all kinds of open IV-accesses in IV-Sets
  • Sterile closing of pre-filled syringes
  • Available in red, blue and white
  • Box of 100 pcs
Specifications
Specifications
Polyethylene
Combi Stopper
Sterile
Blue
Luer Lock