Learn More
B Braun™ Combi-Stopper Closing Cones
B Braun™ Combi-Stopper Closing Cones are suitable for sterile closings of IV and syringe assemblies
£120.00
Specifications
Material | Polyethylene |
---|---|
Product Type | Combi Stopper |
Connection Type | Luer Lock |
Sterility | Sterile |
Description
- Luer Lock fitting male and female
- Made of PE (Polyethylene)
- Sterile closing of all kinds of open IV-accesses in IV-Sets
- Sterile closing of pre-filled syringes
- Available in red, blue and white
- Box of 100 pcs
Specifications
Polyethylene | |
Combi Stopper | |
Luer Lock | |
Sterile |