missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for Easigel H1-SET Horizontal Gel Electrophoresis Unit
For use with Easigel H1-SET Horizontal Gel Electrophoresis Unit
Brand: Fisherbrand™ H1C1512
This item is not returnable.
View return policy
Description
- Color coded
- Offer flexibility in sample-loading
- Volume to maximum 40-sample throughput
Specifications
Easigel H1-SET Horizontal Gel Electrophoresis Unit | |
12 | |
40 μL |
Comb | |
1.5 mm | |
6 mm |