missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for Easigel H1-SET Horizontal Gel Electrophoresis Unit

For use with Easigel H1-SET Horizontal Gel Electrophoresis Unit

Brand:  Fisherbrand™ H1C158

 View more versions of this product

Product Code. 11790545

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Color coded
  • Offer flexibility in sample-loading
  • Volume to maximum 40-sample throughput
Specifications

Specifications

Easigel H1-SET Horizontal Gel Electrophoresis Unit
8
65 μL
Comb
1.5 mm
10 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers