missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for HU6 Mini Horizontal Gel Electrophoresis Unit
For use with HU6 Mini Horizontal Gel Electrophoresis Unit
£55.00 - £92.05
Specifications
For Use With (Equipment) | HU6 Mini Horizontal Gel Electrophoresis Unit |
---|---|
Type | Comb |
Product Code | Brand | Volume (Metric) Sample | Width (Metric) Teeth | No. of Wells | Thickness (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) Sample | Width (Metric) Teeth | No. of Wells | Thickness (Metric) | Price | Quantity & Availability | ||
10707131
|
Fisherbrand™
HU6-C1.5-16 |
15 μL | 2.2 mm | 16 | 1.5 mm | ||||
10090471
|
Fisherbrand™
HU6-C2-12MC |
22 μL | 2.5 mm | 12 | 2 mm | ||||
10738261
|
Fisherbrand™
HU6-C1.5-12MC |
17 μL | 2.5 mm | 12 | 1.5 mm | ||||
10050061
|
Fisherbrand™
HU6-C1.5-8 |
30 μL | 4.5 mm | 8 | 1.5 mm | ||||
11465739
|
Fisherbrand™
HU6-C1-12MC |
11 μL | 2.5 mm | 12 | 1 mm | ||||
10264091
|
Fisherbrand™
HU6-C1-8 |
20 μL | 4.5 mm | 8 | 1 mm | ||||
12395498
|
Fisherbrand™
HU6C216 |
20 μL | 2.2 mm | 16 | 2 mm | ||||
Description
- Color coded
- Height adjustable
- Offer complete control over loading volume
- Well depth to maximum 32-sample throughput
Specifications
HU6 Mini Horizontal Gel Electrophoresis Unit | |
Comb |