Learn More
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 1.5mm Thick
For use in preparatory electrophoresis and high-throughput screening.
£21.90 - £35.40
Specifications
For Use With (Equipment) | MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit |
---|---|
Type | Comb |
Quantity | 1 Each |
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | ||||
11853363
|
Fisherbrand™
MS15-10-1.5 |
10 | 68 μL |
£22.50
Each |
Estimated Shipment: 02-12-2024
Log in to see stock. |
||||
11893363
|
Fisherbrand™
MS15-10MC-1.5 |
10 | 44 μL |
£22.50
Each |
Estimated Shipment: 02-12-2024
Log in to see stock. |
||||
11823373
|
Fisherbrand™
MS15-1-1.5 |
1 Prep, 1 Marker | 743 μL |
£22.50
Each |
Estimated Shipment: 02-12-2024
Log in to see stock. |
||||
11863373
|
Fisherbrand™
MS15-12-1.5 |
12 | 61 μL |
£22.50
Each |
Estimated Shipment: 02-12-2024
Log in to see stock. |
||||
11803383
|
Fisherbrand™
MS15-14MC-1.5 |
14 | 44 μL |
£23.40
Each |
Estimated Shipment: 02-12-2024
Log in to see stock. |
||||
11843383
|
Fisherbrand™
MS15-16MC-1.5 |
16 | 41 μL |
£35.35
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||||
11833393
|
Fisherbrand™
MS15-20-1.5 |
20 | 32 μL |
£35.40
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
||||
11893393
|
Fisherbrand™
MS15-28MCSS-1 |
28 | 11 μL |
£21.90
Each |
Estimated Shipment: 06-11-2024
Log in to see stock. |
||||
11803403
|
Fisherbrand™
MS15-28MCSS-1.5 |
28 | 17 μL |
£35.25
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||||
11843403
|
Fisherbrand™
MS15-30MC-1.5 |
30 | 19 μL |
£35.35
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||||
11883403
|
Fisherbrand™
MS15-35-1.5 |
35 | 15 μL |
£35.35
Each |
Estimated Shipment: 11-11-2024
Log in to see stock. |
||||
11823413
|
Fisherbrand™
MS15-4-1.5 |
4 Prep, 2 Marker | 182 μL |
£22.50
Each |
Estimated Shipment: 02-12-2024
Log in to see stock. |
||||
Description
- Recommended for high resolution gels and publication quality data
- Maximise sample volume
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifications
MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit | |
Comb | |
1 Each |