Learn More
BRAND™ Compact Burette, BLAUBRAND™, Class AS, DE-M, with Stopcock
The New Generation of Burettes from BRAND. With PTFE stopcock, fast to dismantle and easy to clean. Simple repair - all individual components replaceable. Boro 3.3, removable PTFE stopcock. To DIN EN ISO 385. Calibrated to deliver (TD, Ex).
Brand:  BRAND™ 13968
 View more versions of this product
Product Code. 16475979
  • £98.45 / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With removable PTFE stopcock, fast to dismantle and easy to clean
  • Simple repair - all individual components replaceable
  • To DIN EN ISO 385
  • Calibrated to deliver (TD, Ex)
All BLAUBRAND™ burettes are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Amber
Borosilicate Glass (3.3)
Burette tube, stopcock with precision tip, one batch certificate
Compact Buret
50 mL
0.1 mL
PTFE
Class AS