Learn More
Nabertherm™ Compact Tube Furnace with P580 Controller
These compact tube furnaces with integrated control systems can be used universally for many processes. Equipped with a standard working tube of C 530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.
£5280.00 - £10295.00
Specifications
Controller Type P580
Type Compact Tube Furnace
View More Specs
Products 7
Product Code Brand Diameter (Metric) Process Tube Heated Zone Temperature (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Process Tube Heated Zone Temperature (Metric) Price Quantity & Availability  
17398079
View Documents
Nabertherm™
R-252W2AN3
50 mm 500 mm 1200°C
£5280.00
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
17308089
View Documents
Nabertherm™
R-252W3AN3
50 mm 500 mm 1300°C
£5715.00
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
17358089
View Documents
Nabertherm™
R-322W2AN3
120 mm 500 mm 1200°C
£6285.00
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
17368089
View Documents
Nabertherm™
R-322W3AN3
120 mm 500 mm 1300°C
£6975.00
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
17318099
View Documents
Nabertherm™
R-472W2AN3
170 mm 750 mm 1200°C
£8915.00
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
17328099
View Documents
Nabertherm™
R-472W3AN3
170 mm 750 mm 1300°C
£9925.00
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
17378099
View Documents
Nabertherm™
R-572W2AN3
170 mm 1000 mm 1200°C
£10295.00
Each
Estimated Shipment: 23-05-2025
to see stock.
Add to basket
Description
Description
  • Excellent price - performance ratio
  • Can also be equipped with gas supply system 1 (no vacuum operation) and gas supply system 2 (for vacuum operation)
  • Temperature, max. 1200°C or 1300°C
  • Controller P580 with 50 freely adjustable programs and 40 segments for three-zone operation
  • Outer tube diameter of 50 to 170 mm, heated length from 250 to 1000 mm
  • Dual shell housing made of sheets of textured stainless steel
  • Insulation made of non-classified fibre material
Specifications
Specifications
P580
Compact Tube Furnace
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Nabertherm™ Compact Tube Furnace with P580 Controller