Learn More
Agilent BioTek Complete Direct Drain Waste Systems
Drains into sink instead of bottles
Brand:  Agilent BioTek 1170560-S
 View more versions of this product
Product Code. 15447180
  • £6189.00 / Each
Explore more special offers
Description
Description
  • Compatible with Agilent BioTek 405 and 406 Series Microplate Washer Systems
  • Eliminates the need of attending to and emptying waste bottles
  • Saves space on the workbench
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Direct drain module (115V/230V), standard or high-flow vacuum pump, tubing, holder for two 4L bottles
Complete Direct Drain Waste System
Agilent BioTek 405 Series Microplate Washer System, Agilent BioTek 406 Series Microplate Washer System