Learn More
Agilent BioTek Complete Direct Drain Waste Systems
GREENER_CHOICE
Drains into sink instead of bottles
£6189.00 - £11202.00
Specifications
Includes Direct drain module (115V/230V), standard or high-flow vacuum pump, tubing, holder for two 4L bottles
For Use With (Equipment) Agilent BioTek 405 Series Microplate Washer System, Agilent BioTek 406 Series Microplate Washer System
View More Specs
Products 2
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
15447180
View Documents
Agilent BioTek
1170560-S
Complete Direct Drain Waste System
£6189.00
Each
15993300
GREENER_CHOICE
View Documents
Agilent BioTek
1170562-S
£11202.00
Each
Description
Description
  • Compatible with Agilent BioTek 405 and 406 Series Microplate Washer Systems
  • Eliminates the need of attending to and emptying waste bottles
  • Saves space on the workbench
Specifications
Specifications
Direct drain module (115V/230V), standard or high-flow vacuum pump, tubing, holder for two 4L bottles
Agilent BioTek 405 Series Microplate Washer System, Agilent BioTek 406 Series Microplate Washer System