missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Con A Sepharose™ 4B Medium
An affinity chromatography media for capture of glycosylated biomolecules including glycoproteins and glycolipids
£100.29 - £834.08
Specifications
Binding Capacity | 30mg/mL |
---|---|
Ligand Concentration | 13mg Con A/mL Drained Medium |
Matrix | 4% Agarose |
Particle Size | 90 μm |
pH | 4 to 9 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11534905
|
Cytiva
17-0440-03 |
5 mL |
£100.29
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11524905
|
Cytiva
17-0440-01 |
100 mL |
£834.08
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Highly suited for isolation of cell surface glycoproteins from detergent-solubilized membranes.
- Con A coupled to Sepharose, in a 4% agarose matrix via cyanogen bromide activation
- Binds molecules that contain α-D-mannose, α-D-glucose, and sterically related residues with available C-3, C-4, or C-5 hydroxyl groups
- Group-specific adsorbent for molecules containing sugars
- Concentration of immobilized Con A, approx. 13mg/mL medium; binding capacity for porcine thyroglobulin, approx. 30mg/mL medium
- Stable in the pH range 4 to 9, and in the presence of all commonly used buffers
- Average particle size: 90μm
- Delivered in 0.1M acetate buffer, pH 6.0, containing 1M NaCl, 1mM CaCl2, 1mM MgCl2, 1mM MnCl2, and 20% ethanol
Specifications
30mg/mL | |
4% Agarose | |
4 to 9 | |
For separation and purification of Glycoproteins, Polysaccharides and Glycolipids, isolates cell surface Glycoproteins from detergent-solubilized membranes |
13mg Con A/mL Drained Medium | |
90 μm | |
2°C to 8°C | |
Concanavalin A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Con A Sepharose™ 4B Medium