Learn More
VELP Scientifica™ Condensation Kit for Water Samples
The condensation kit for water samples consists of a 3.5 liters tank and a condenser that permits the condensation and collection of the evaporated liquid without requiring any operator intervention.
Brand:  VELP Scientifica™ A00000477
Product Code. 17824687
  • £469.00 / Each
Estimated Shipment: 29-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The condensation kit permits the condensation and collection of the evaporated liquid without operator intervention.
  • Consists of a 3.5 liter tank
  • For low nitrogen content samples, such as water samples, the TKN analysis requires a high sample volume.
  • During the digestion step, the evaporation of the liquid could lead to the filling of the KS 1000 dilution tank and a relevant increase in the reagents’ temperature.
  • Required when the sum of the sample volumes to be digested is more than 500 mL. 
  • Can be used in substituion of the dilution tank or it can be used as an additional stage.
Specifications
Specifications
Condensation Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
VELP Scientifica™ Condensation Kit for Water Samples > Velp Scientifica™ Consendation Kit