Learn More
Velp Scientifica™ Test Tubes for Digestion Units
Facilitates the nitrogen analysis and protein determination process. Velp Scientifica™ Test Tubes for Digestion Units are for use with Velp Scientifica™ Kjeldahl Digestion Units.
Brand:  Velp Scientifica™ A00000144
 View more versions of this product
Product Code. 11715654
  • £81.95 / Pack of 3
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use in Total Kjeldahl Nitrogen Determination, ammoniacal nitrogen, nitric nitrogen (Devarda), TVBN, sulphites, phenols, volatile acids and cyanides
Specifications
Specifications
Test Tubes
42 mm
3 Tubes
250 mL
300 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Brochures
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Velp Scientifica™ Test Tubes for Digestion Units > Nitrogen Determination Test Tubes; Capacity (Metric): 250 mL; Diameter (Metric) Outer: 42 mm; Length (Metric): 300 mm; Product Type: Test Tubes