Learn More
Lenz Laborglasintrumente Condensers acc. to Allihn
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05223312
 View more versions of this product
Product Code. 11973421
  • £81.80 / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Allihn condensers acc. to DIN 12581
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: GL 14 threads (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifications
Specifications
Two threaded plastic olives, GL threads 14
250 mm
NS 19/26