Learn More
Lenz Laborglasintrumente Condensers Acc. to Davies
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05215404
Additional Details : Weight : 5.00000kg
Description
- Davies condensers for condensing vapours
- With NS socket and NS cone
- With double sized cooling area
- Water connection: glass olives
- Made of DURAN™ tubing
Specifications
Glass olives | |
NS 29/32 | |
Borosilicate Glass (3.3) | |
400 mm |