Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig, with GL Threads
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05210311
 View more versions of this product
Product Code. 11526794
  • £42.45 / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Liebig condensers acc. to DIN 12576 and in addition to DIN
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: GL 14 threads (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifications
Specifications
GL 14 Threads
250 mm
NS 14/23