Learn More
Lenz Laborglasintrumente Condensers Acc. to Liebig with GL Threads
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05210413
Additional Details : Weight : 0.50000kg
Description
- Liebig condensers acc. to DIN 12576 and in addition to DIN
- For condensing vapours
- With NS socket and NS cone
- Water connection: GL 14 threads (with two threaded plastic olives)
- Made of DURAN™ tubing
Specifications
GL 14 threads with two threaded plastic olives | |
400 mm | |
400 mm |
NS 24/29 | |
1 Ea. |