missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jenway™ Bench pH/Conductivity Meter 3540
Ideal for use in all laboratories where pH and conductivity analyses are required. The setup menu gives quick and easy access to the whole range of instrument, pH and conductivity measurement options.
Brand: Jenway™ 99959-96
1558.84 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
The 3540 conductivity pH meter is supplied with a glass combination pH electrode (924 005), glass conductivity probe (K=1 027 013), electrode stand and holder, ATC probe (027 500), BNC shorting plug, pH buffers and UK power supply.Specifications
Ideal for use in all laboratories where pH and conductivity analyses are required | |
Calibration of both channels is automatic, with the option to select 1, 2 or 3 calibration points. | |
Storage of up to 500 readings (250 for each mode) | |
pH/Conductivity Meter |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction