Learn More
Hanna Instruments™ Conductivity solution HI-7034 c/w certificate of analysis 80,000μS/cm (μmho/cm) 460mL Hanna
Brand:  Hanna Instruments™ HI7034+COA
Product Code. 11504337
  • £130.00 / Each
Explore more special offers
Description
Description
A choice of ready mixed calibration standards traceable to NIST. Available in 460mL bottles or 20mL individually sealed sachets to protect against light and air. Supplied in boxes of 25.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hanna Instruments™ Conductivity solution HI-7034 c/w certificate of analysis 80,000μS/cm (μmho/cm) 460mL Hanna >