Learn More
Emma Safety Footwear Constans Safety Shoes
Developed for semiorthopedic adjustments requiring customized in-soles.
Brand: Emma Safety Footwear MM13207040
Additional Details : Weight : 0.50000kg
Description
- Full grain leather upper construction
- Nappa leather lining
- With sole plate for wet work
- Woven zero penetration in-sole
- Antibacterial inlay sole
- Vibram™ Nitril / HRO outsole
Specifications
Black | |
Nappa leather | |
40 | |
Vibram™ Nitrile |
semiorthopedic adjustments requiring customized in-soles | |
Leather | |
Woven zero-penetration insole, Heat-resistant outsole | |
Constans Safety Shoes |